Läs mer
N-Pivaloylglycine, TRC
Produktkod. 30420535
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30420535 50 mg
1 options
Produktkod. 30420535 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP55074050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Pivaloylglycine
CAS 23891-96-5
Molekylformel C7H13NO3
Kvantitet 50 mg
Synonym N-(2,2-Dimethyl-1-oxopropyl)-glycine, N-Pivaloyl-glycine
InChI formel InChI=1S/C7H13NO3/c1-7(2,3)6(11)8-4-5(9)10/h4H2,1-3H3,(H,8,11)(H,9,10)
LEDER CC(C)(C)C(=O)NCC(=O)O
IUPAC-namn 2-(2,2-dimethylpropanoylamino)acetic acid
Molekylvikt (g/mol) 159.18
Formel vikt 159.0895