Läs mer
N-Propionyl-L-cysteine, Mikromol™
Produktkod. 30049195
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30049195 25 mg
30050260 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30049195 Leverantör Mikromol Leverantörsnummer MM0024.060025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N-Propionyl-L-cysteine
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Propionyl-L-cysteine
CAS 2885-79-2
Analyt- eller komponentnamn N-Propionyl-L-cysteine
Frakt skick Room Temperature
Molekylformel C6 H11 N O3 S
Certifikat och efterlevnad ISO 9001
Synonym N-Propionyl-L-cysteine
InChI formel InChI=1S/C6H11NO3S/c1-2-5(8)7-4(3-11)6(9)10/h4,11H,2-3H2,1H3,(H,7,8)(H,9,10)/t4-/m0/s1
LEDER CCC(=O)N[C@@H](CS)C(=O)O
IUPAC-namn (2R)-2-(propanoylamino)-3-sulfanylpropanoic acid
Molekylvikt (g/mol) 177.22
Kvantitet 25 mg
Formel vikt 177.046 g/mol