Läs mer
N-(Propyl(2-(2,4,6-trichlorophenoxy)ethyl)carbamoyl)formamide, TRC
Produktkod. 17981485
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17981485 100 mg
17973945 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17981485 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP838880100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 139542-32-8
Molekylformel C13 H15 Cl3 N2 O3
Molekylvikt (g/mol) 353.63
Synonym Urea, N'-formyl-N-propyl-N-[2-(2,4,6-trichlorophenoxy)ethyl]-, Prochloraz desimidazole-formylamino, N-(Propyl(2-(2,4,6-trichlorophenoxy)ethyl)carbamoyl)formamide, BTS 44596
IUPAC-namn N-[propyl-[2-(2,4,6-trichlorophenoxy)ethyl]carbamoyl]formamide
LEDER CCCN(CCOc1c(Cl)cc(Cl)cc1Cl)C(=O)NC=O
Specifikationer
Kemiskt namn eller material N-(Propyl(2-(2,4,6-trichlorophenoxy)ethyl)carbamoyl)formamide
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C13H15Cl3N2O3/c1-2-3-18(13(20)17-8-19)4-5-21-12-10(15)6-9(14)7-11(12)16/h6-8H,2-5H2,1H3,(H,17,19,20)
Formel vikt 352.0148
Procent renhet >95%