Läs mer
n-Propylcyclopentane, TRC
Produktkod. 30418305
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30418305 25 mg
1 options
Produktkod. 30418305 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP83516025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material n-Propylcyclopentane
CAS 2040-96-2
Molekylformel C8 H16
Kvantitet 25 mg
Synonym Cyclopentane','propyl-, Propylcyclopentane, 1-Propylcyclopentane, NSC 73947, n-Propylcyclopentane
InChI formel InChI=1S/C8H16/c1-2-5-8-6-3-4-7-8/h8H,2-7H2,1H3
LEDER CCCC1CCCC1
IUPAC-namn propylcyclopentane
Molekylvikt (g/mol) 112.21
Formel vikt 112.1252