Läs mer
N-Pyrazinylcarbonyl-L-phenylalanine Methyl Ester, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | N-Pyrazinylcarbonylphenylalanine Methyl Ester |
| CAS | 73058-37-4 |
| Molekylformel | C15 H15 N3 O3 |
| Kvantitet | 100 mg |
| Synonym | L-Phenylalanine','N-(2-pyrazinylcarbonyl)-','methyl ester (ACI), L-Phenylalanine','N-(pyrazinylcarbonyl)-','methyl ester (9CI), N-(2-Pyrazinylcarbonyl)-L-phenylalanine methyl ester (ACI), Methyl (S)-3-phenyl-2-[[(pyrazin-2-yl)carbonyl]amino]propanoate |
| InChI formel | InChI=1S/C15H15N3O3/c1-21-15(20)12(9-11-5-3-2-4-6-11)18-14(19)13-10-16-7-8-17-13/h2-8,10,12H,9H2,1H3,(H,18,19) |
| LEDER | COC(=O)C(Cc1ccccc1)NC(=O)c2cnccn2 |
| IUPAC-namn | methyl 3-phenyl-2-(pyrazine-2-carbonylamino)propanoate |
| Molekylvikt (g/mol) | 285.3 |
| Formel vikt | 285.1113 |