Läs mer
N-[(S)-(4-Nitrophenoxy)phenoxyphosphinyl]-L-alanine 1-Methylethyl Ester, TRC
Produktkod. 30482364
Change view
Kvantitet:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30482364 25 mg
30466746 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30482364 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN49703025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-[(S)-(4-Nitrophenoxy)phenoxyphosphinyl]-L-alanine 1-Methylethyl Ester
CAS 1256490-31-9
Molekylformel C15 H14 O2
Kvantitet 25 mg
Synonym Remdesivir Impurity 25
InChI formel InChI=1S/C15H14O2/c1-11-13(12-7-4-3-5-8-12)9-6-10-14(11)15(16)17-2/h3-10H,1-2H3
LEDER COC(=O)c1cccc(c1C)c2ccccc2
IUPAC-namn methyl 2-methyl-3-phenylbenzoate
Molekylvikt (g/mol) 226.27
Formel vikt 226.099