Läs mer
N,S-Diacetyl-L-cysteine, TRC
Produktkod. 17914505
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17914505 50 mg
17901645 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17914505 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD31230050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 18725-37-6
Molekylformel C7 H11 N O4 S
Molekylvikt (g/mol) 205.23
Synonym N,S-Diacetyl-L-cysteine, Acetylcysteine Imp. D (EP)
IUPAC-namn (2R)-2-acetamido-3-acetylsulfanylpropanoic acid
LEDER CC(=O)N[C@@H](CSC(=O)C)C(=O)O
Specifikationer
Kemiskt namn eller material N,S-Diacetyl-L-cysteine
Anteckningar om renhetsgrad HPLC
InChI formel InChI=1S/C7H11NO4S/c1-4(9)8-6(7(11)12)3-13-5(2)10/h6H,3H2,1-2H3,(H,8,9)(H,11,12)/t6-/m0/s1
Kvantitet 50 mg
Formel vikt 205.0409
Procent renhet >95%