Läs mer
N-(S)-Glycidylphthalimide, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | N-(S)-Glycidylphthalimide |
| CAS | 161596-47-0 |
| Molekylformel | C11H9NO3 |
| Kvantitet | 1 g |
| Synonym | 1H-Isoindole-1,3(2H)-dione, 2-[(2S)-2-oxiranylmethyl]- (ACI), 1H-Isoindole-1,3(2H)-dione, 2-(oxiranylmethyl)-, (S)- (ZCI), 1H-Isoindole-1,3(2H)-dione, 2-[(2S)-oxiranylmethyl]- (9CI), 2-[(2S)-2-Oxiranylmethyl]-1H-isoindole-1,3(2H)-dione (ACI), (S)-2-(Oxiran-2-ylmethyl)-1H-isoindole-1,3(2H)-dione, (S)-2-(Oxiran-2-ylmethyl)isoindoline-1,3-dione, (S)-2-[(Oxiranyl)methyl]isoindole-1,3-dione, (S)-Glycidyl Phthalimide, (S)-N-(2,3-Epoxypropan-1-yl)phthalimide, (S)-N-(2,3-Epoxypropyl)phthalimide, (S)-N-Giycidylphthalimide, (S)-N-Glycidylphthalimide, 2-[(S)-2-Oxiranylmethyl]-1H-isoindol-1,3(2H)-dione, 2-[[(2S)-Oxiran-2-yl]methyl]isoindole-1,3-dione, N-((2S)-Oxiran-2-ylmethyl)phthalimide, N-(S)-Glycidylphthalimide, N-[(+)-Glycidyl]phthalimide, N-[(S)-(+)-Glycidyl]phthalimide, (S)-2-(Oxiran-2-ylmethyl)-1H-isoindole-1,3(2H)-dione, (S)-2-[(Oxiranyl)methyl]isoindole-1,3-dione, (S)-N-(2,3-Epoxypropan-1-yl)phthalimide, (S)-N-(2,3-Epoxypropyl)phthalimide, 2-[(S)-2-Oxiranylmethyl]-1H-isoindol-1,3(2H)-dione, N-((2S)-Oxiran-2-ylmethyl)phthalimide |
| InChI formel | InChI=1S/C11H9NO3/c13-10-8-3-1-2-4-9(8)11(14)12(10)5-7-6-15-7/h1-4,7H,5-6H2/t7-/m0/s1 |
| LEDER | O=C1N(C[C@H]2CO2)C(=O)c3ccccc13 |
| IUPAC-namn | 2-[[(2S)-oxiran-2-yl]methyl]isoindole-1,3-dione |
| Molekylvikt (g/mol) | 203.19 |
| Formel vikt | 203.0582 |