Läs mer
N-Succinimidoxycarbonyl-Beta-alanine N-Succinimidyl Ester 1,4- Dioxane complex, TRC
Produktkod. 30418435
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30418435 250 mg
1 options
Produktkod. 30418435 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS689850250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Succinimidoxycarbonyl-beta-alanine N-Succinimidyl Ester','1,4-Dioxane
CAS 1215667-16-5
Molekylformel C12 H13 N3 O8 . C4 H8 O2
Kvantitet 250 mg
Synonym N-Succinimidoxycarbonyl-B-Alanine N-Succinimidyl Ester 1,4- Dioxane Complex
InChI formel InChI=1S/C12H13N3O8.C4H8O2/c16-7-1-2-8(17)14(7)22-11(20)5-6-13-12(21)23-15-9(18)3-4-10(15)19;1-2-6-4-3-5-1/h1-6H2,(H,13,21);1-4H2
LEDER O=C(CCNC(=O)ON1C(=O)CCC1=O)ON2C(=O)CCC2=O.C3COCCO3
IUPAC-namn 1,4-dioxane;(2,5-dioxopyrrolidin-1-yl) 3-[(2,5-dioxopyrrolidin-1-yl)oxycarbonylamino]propanoate
Molekylvikt (g/mol) 415.35
Formel vikt 415.1227