Läs mer
N-Succinimidyl 3-Trimethylstannyl-benzoate, TRC
Produktkod. 30135036
Change view
Kvantitet:
2.5 mg
5 mg
25 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30135036 25 mg -
30133935 2.5 mg 2.5mg
17983825 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30135036 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS69202525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 122856-01-3
Molekylformel C14 H17 N O4 Sn
Molekylvikt (g/mol) 382
IUPAC-namn (2,5-dioxopyrrolidin-1-yl) 3-trimethylstannylbenzoate
LEDER C[Sn](C)(C)c1cccc(c1)C(=O)ON2C(=O)CCC2=O
Specifikationer
Kemiskt namn eller material N-Succinimidyl 3-Trimethylstannyl-benzoate
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C11H8NO4.3CH3.Sn/c13-9-6-7-10(14)12(9)16-11(15)8-4-2-1-3-5-8;;;;/h1-2,4-5H,6-7H2;3*1H3;
Formel vikt 383.018
Procent renhet >95%