Läs mer
N-t-Boc-L-serine Allyl Ester, TRC
Produktkod. 30345856
Change view
Kvantitet:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30345856 50 mg
30336822 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30345856 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB66737550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-t-Boc-L-serine Allyl Ester
CAS 143966-57-8
Molekylformel C11 H19 N O5
Kvantitet 50 mg
Synonym L-Serine, N-[(1,1-dimethylethoxy)carbonyl]-, 2-propen-1-yl ester, L-Serine, N-[(1,1-dimethylethoxy)carbonyl]-, 2-propenyl ester (9CI), (S)-Allyl 2-((tert-butoxycarbonyl)amino)-3-hydroxypropanoate, N-(tert-Butoxycarbonyl)serine allyl ester, N-(tert-Butoxycarbonyl)-L-serine allyl ester, N-t-Boc-L-serine allyl ester
InChI formel InChI=1S/C11H19NO5/c1-5-6-16-9(14)8(7-13)12-10(15)17-11(2,3)4/h5,8,13H,1,6-7H2,2-4H3,(H,12,15)/t8-/m0/s1
LEDER CC(C)(C)OC(=O)N[C@@H](CO)C(=O)OCC=C
IUPAC-namn prop-2-enyl (2S)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
Molekylvikt (g/mol) 245.27
Formel vikt 245.1263