Learn More
N-(tert-Butoxycarbonyl)-D-prolinol, TRC
Specifications
Specifications
| Kemiskt namn eller material | N-(tert-Butoxycarbonyl)-D-prolinol |
| CAS | 83435-58-9 |
| Molekylformel | C10H19NO3 |
| Kvantitet | 100 g |
| Synonym | (2R)-(+)-1-Boc-2-pyrrolidinemethanol, (R)-(+)-1-(tert-Butoxycarbonyl)-2-pyrrolidinemethanol, (R)-(+)-2-Hydroxymethylpyrrolidine-1-carboxylic acid tert-butyl ester, (R)-1-(tert-Butoxycarbonyl)-2-(hydroxymethyl)pyrrolidine, (R)-1-(tert-Butoxycarbonyl)-2-pyrrolidinemethanol, (R)-1-Boc-2-pyrrolidinemethanol, (R)-1-tert-Butyloxycarbonyl-2-(hydroxymethyl)pyrrolidine, (R)-2-(Hydroxymethyl)pyrrolidin-1-carboxylic acid tert-butyl ester, (R)-2-(Hydroxymethyl)pyrrolidine-1-carboxylic acid tert-butyl ester, (R)-2-Hydroxymethylpyrrolidine-1-carboxylic acid tert-butyl ester, (R)-[1-(tert-Butoxycarbonyl)pyrrolidin-2-yl]methanol, (R)-tert-Butyl 2-(hydroxymethyl)pyrrolidine-1-carboxylate, 1,1-Dimethylethyl (2R)-2-(hydroxymethyl)-1-pyrrolidinecarboxylate, 1-BOC-2-(R)-pyrrolidinemethanol, 2-(Hydroxymethyl)pyrrolidine-1-carboxylic acid (R)-tert-butyl ester, N-Boc-D-prolinol, tert-Butyl (2R)-2-(hydroxymethyl)pyrrolidine-1-carboxylate, tert-Butyl (2R)-2-(hydroxymethyl)pyrrolidine-1-carboxylate, tert-Butyl (R)-2-(hydroxymethyl)-1-pyrrolidinecarboxylate |
| InChI formel | InChI=1S/C10H19NO3/c1-10(2,3)14-9(13)11-6-4-5-8(11)7-12/h8,12H,4-7H2,1-3H3/t8-/m1/s1 |
| LEDER | OC[C@@H]1N(C(OC(C)(C)C)=O)CCC1 |
| IUPAC-namn | tert-butyl (R)-2-(hydroxymethyl)pyrrolidine-1-carboxylate |
| Molekylvikt (g/mol) | 201.26 |
| Formel vikt | 201.14 |