Läs mer
N-(tert-Butoxycarbonyl)-L-cysteine Methyl Ester, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | N-(tert-Butoxycarbonyl)-L-cysteine Methyl Ester |
| CAS | 55757-46-5 |
| Molekylformel | C9H17NO4S |
| Kvantitet | 5 g |
| Synonym | N-[(1,1-Dimethylethoxy)carbonyl]-L-cysteine Methyl Ester, Boc-L-cysteine Methyl Ester, Methyl (tert-Butoxycarbonyl)-L-cysteinate, N-Boc-L-cysteine Methyl Ester |
| InChI formel | InChI=1S/C9H17NO4S/c1-9(2,3)14-8(12)10-6(5-15)7(11)13-4/h6,15H,5H2,1-4H3,(H,10,12)/t6-/m0/s1 |
| LEDER | COC(=O)[C@H](CS)NC(=O)OC(C)(C)C |
| IUPAC-namn | methyl (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-sulfanylpropanoate |
| Molekylvikt (g/mol) | 235.3 |
| Formel vikt | 235.0878 |