Läs mer
N-tert-Butyl-N-ethylnitrosamine, TRC
Produktkod. 30329710
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30329710 50 mg
1 options
Produktkod. 30329710 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB81024050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-tert-Butyl-N-ethylnitrosamine
CAS 3398-69-4
Molekylformel C6 H14 N2 O
Kvantitet 50 mg
Synonym N-Ethyl-2-methyl-N-nitroso-2-propanamine, Ethyl-tert-butylnitrosamine, N-Ethyl-N-tert-butylnitrosamine, N-Nitroso-N-ethyl-tert-butylamine
InChI formel InChI=1S/C6H14N2O/c1-5-8(7-9)6(2,3)4/h5H2,1-4H3
LEDER CCN(N=O)C(C)(C)C
IUPAC-namn N-tert-butyl-N-ethyl-nitrous amide
Molekylvikt (g/mol) 130.19
Formel vikt 130.1106