Läs mer
N-tert-Butyloxycarbonyl-L-aspartic Acid b-tert-Butyl Ester, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | N-tert-Butyloxycarbonyl-L-aspartic Acid beta-tert-Butyl Ester |
| CAS | 1676-90-0 |
| Molekylformel | C13H23NO6 |
| Kvantitet | 5 g |
| Synonym | tert-Butyloxycarbonyl-L-aspartic Acid 4-tert-Butyl Ester, L-N-Carboxyaspartic Acid N,4-di-tert-Butyl Ester, (S)-4-tert-Butoxy-2-[(tert-butoxycarbonyl)amino]-4-oxobutanoic Acid, 4-tert-Butyl (S)-2-tert-Butoxycarbonylaminosuccinate, BOC-L-Asp(Obut)-OH, Boc-L-Asp(OtBu)-OH, N-(tert-Butoxycarbonyl)-L-aspartic Acid beta-tert-Butyl Ester, N-Boc-L-asparaginic Acid gamma-(tert-Butyl) Ester, N-Boc-L-aspartic Acid beta-tert-Butyl Ester, N-tert-Butoxycarbonylaspartic Acid gamma-tert-Butyl Ester |
| InChI formel | InChI=1S/C13H23NO6/c1-12(2,3)19-9(15)7-8(10(16)17)14-11(18)20-13(4,5)6/h8H,7H2,1-6H3,(H,14,18)(H,16,17)/t8-/m0/s1 |
| LEDER | CC(C)(C)OC(=O)C[C@H](NC(=O)OC(C)(C)C)C(=O)O |
| IUPAC-namn | (2S)-4-[(2-methylpropan-2-yl)oxy]-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid |
| Molekylvikt (g/mol) | 289.32 |
| Formel vikt | 289.1525 |