Läs mer
N-Tosyl-L-alanine, TRC
Produktkod. 30453896
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30453896 500 mg
1 options
Produktkod. 30453896 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT550525500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Tosyl-L-alanine
CAS 21957-58-4
Molekylformel C10 H13 N O4 S
Kvantitet 500 mg
Synonym L-Alanine','N-[(4-methylphenyl)sulfonyl]-, Alanine','N-(p-tolylsulfonyl)-','L- (8CI), N-[(4-Methylphenyl)sulfonyl]-L-alanine, (2S)-2-[[(4-Methylphenyl)sulfonyl]amino]propanoic acid, (S)-2-((4-Methylphenylsulfonyl)amino)propionic acid, N-(Toluene-4-sulfonyl)-L-alanine, N-(p-Tolylsulfonyl)alanine, N-Tosyl-L-alanine, N-Tosylalanine, N-p-Tolylsulfonyl-L-alanine, Ts-L-Ala-OH, p-Toluenesulfonyl-L-alanine
InChI formel InChI=1S/C10H13NO4S/c1-7-3-5-9(6-4-7)16(14,15)11-8(2)10(12)13/h3-6,8,11H,1-2H3,(H,12,13)/t8-/m0/s1
LEDER C[C@H](NS(=O)(=O)c1ccc(C)cc1)C(=O)O
IUPAC-namn (2S)-2-[(4-methylphenyl)sulfonylamino]propanoic acid
Molekylvikt (g/mol) 243.28
Formel vikt 243.0565