Läs mer
N-Tosyl-L-glutamic Acid, TRC
Produktkod. 30451405
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30451405 1 g
1 options
Produktkod. 30451405 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT6649851G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Tosyl-L-glutamic Acid
CAS 4816-80-2
Molekylformel C12H15NO6S
Kvantitet 1 g
Synonym N-p-Tolylsulfonyl-L-glutamic Acid, (+)-p-Toluenesulfonylglutamic Acid, Tosylglutamic Acid, NSC 109186
InChI formel InChI=1S/C12H15NO6S/c1-8-2-4-9(5-3-8)20(18,19)13-10(12(16)17)6-7-11(14)15/h2-5,10,13H,6-7H2,1H3,(H,14,15)(H,16,17)/t10-/m0/s1
LEDER Cc1ccc(cc1)S(=O)(=O)N[C@@H](CCC(=O)O)C(=O)O
IUPAC-namn (2S)-2-[(4-methylphenyl)sulfonylamino]pentanedioic acid
Molekylvikt (g/mol) 301.32
Formel vikt 301.062