Läs mer
N-(Trifluoroacetyl)hexanolamine, TRC
Produktkod. 30456060
Change view
Kvantitet:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30456060 1 g
30453810 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30456060 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT7880501G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 40248-34-8
Molekylformel C8 H14 F3 N O2
Molekylvikt (g/mol) 213.2
Synonym 6-(Trifluoroacetamido)-1-hexanol, 6-(Trifluoroacetamido)hexanol, 6-(Trifluoroacetylamino)-1-hexanol, 6-[(Trifluoroacetyl)amino]hexanol, N-(Trifluoroacetyl)-6-amino-1-hexanol
IUPAC-namn 2,2,2-trifluoro-N-(6-hydroxyhexyl)acetamide
LEDER OCCCCCCNC(=O)C(F)(F)F
Specifikationer
Kemiskt namn eller material N-(Trifluoroacetyl)hexanolamine
Kvantitet 1 g
InChI formel InChI=1S/C8H14F3NO2/c9-8(10,11)7(14)12-5-3-1-2-4-6-13/h13H,1-6H2,(H,12,14)
Formel vikt 213.0977