Läs mer
N-Trityl Candesartan Ethyl Ester, TRC
Produktkod. 30456019
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30456019 5 mg
1 options
Produktkod. 30456019 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT8872105MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Trityl Candesartan Ethyl Ester
CAS 1797985-81-9
Molekylformel C45H38N6O3
Kvantitet 5 mg
Synonym 2-Ethoxy-1-[[2'-[2-(triphenylmethyl)-2H-tetrazol-5-yl][1,1'-biphenyl]-4-yl]methyl]-1H-benzimidazole-7-carboxylic Acid Ethyl Ester
InChI formel InChI=1S/C45H38N6O3/c1-3-53-43(52)39-25-16-26-40-41(39)50(44(46-40)54-4-2)31-32-27-29-33(30-28-32)37-23-14-15-24-38(37)42-47-49-51(48-42)45(34-17-8-5-9-18-34,35-19-10-6-11-20-35)36-21-12-7-13-22-36/h5-30H,3-4,31H2,1-2H3
LEDER CCOC1=NC2=CC=CC(=C2N1CC3=CC=C(C=C3)C4=CC=CC=C4C5=NN(N=N5)C(C6=CC=CC=C6)(C7=CC=CC=C7)C8=CC=CC=C8)C(=O)OCC
Molekylvikt (g/mol) 710.82
Formel vikt 710.3005