Läs mer
N-Trityl-deshydroxymethyl Losartan, TRC
Produktkod. 30454625
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30454625 1 g
30453991 500 mg
30456510 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30454625 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT8878251G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1797133-13-1
Molekylformel C40H35ClN6
Molekylvikt (g/mol) 635.21
Synonym 5-[4'-[(2-Butyl-4-chloro-1H-imidazol-1-yl)methyl][1,1'-biphenyl]-2-yl]-2-(triphenylmethyl)-2H-tetrazole
IUPAC-namn 5-[2-[4-[(2-butyl-4-chloroimidazol-1-yl)methyl]phenyl]phenyl]-2-trityltetrazole
LEDER CCCCc1nc(Cl)cn1Cc2ccc(cc2)c3ccccc3c4nnn(n4)C(c5ccccc5)(c6ccccc6)c7ccccc7
Specifikationer
Kemiskt namn eller material N-Trityl-deshydroxymethyl Losartan
Kvantitet 1 g
InChI formel InChI=1S/C40H35ClN6/c1-2-3-23-38-42-37(41)29-46(38)28-30-24-26-31(27-25-30)35-21-13-14-22-36(35)39-43-45-47(44-39)40(32-15-7-4-8-16-32,33-17-9-5-10-18-33)34-19-11-6-12-20-34/h4-22,24-27,29H,2-3,23,28H2,1H3
Formel vikt 634.2612