Läs mer
N-Trityl Losartan Isomer, TRC
Produktkod. 30450364
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30450364 250 mg
30452317 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30450364 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT888835250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 150098-04-7
Molekylformel C41H37ClN6O
Molekylvikt (g/mol) 665.23
Synonym 2-Butyl-5-chloro-1-[[2'-[2-(triphenylmethyl)-2H-tetrazol-5-yl][1,1'-biphenyl]-4-yl]methyl]-1H-imidazole-4-methanol
IUPAC-namn [2-butyl-5-chloro-1-[[4-[2-(2-trityltetrazol-5-yl)phenyl]phenyl]methyl]imidazol-4-yl]methanol
LEDER CCCCc1nc(CO)c(Cl)n1Cc2ccc(cc2)c3ccccc3c4nnn(n4)C(c5ccccc5)(c6ccccc6)c7ccccc7
Specifikationer
Kemiskt namn eller material N-Trityl Losartan Isomer
Kvantitet 250 mg
InChI formel InChI=1S/C41H37ClN6O/c1-2-3-23-38-43-37(29-49)39(42)47(38)28-30-24-26-31(27-25-30)35-21-13-14-22-36(35)40-44-46-48(45-40)41(32-15-7-4-8-16-32,33-17-9-5-10-18-33)34-19-11-6-12-20-34/h4-22,24-27,49H,2-3,23,28-29H2,1H3
Formel vikt 664.2717