missing translation for 'onlineSavingsMsg'
Läs mer

N-Tritylolmesartan Ethyl Ester (4-(1-Hydroxy-1-methylethyl)-2-propyl-1-[[2'-[2-(triphenylmethyl)-2H-tetrazol-5-yl][1,1'-biphenyl]-4-yl]methyl]-1H-imidazole-5-carboxylic Acid Ethyl Ester), Mikromol™

Product Code. 18081680
Change view
Click to view available options
Kvantitet:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
18081680 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18081680 Supplier Mikromol Supplier No. MM1317.280025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

N-Tritylolmesartan Ethyl Ester (4-(1-Hydroxy-1-methylethyl)-2-propyl-1-[[2'-[2-(triphenylmethyl)-2H-tetrazol-5-yl][1,1'-biphenyl]-4-yl]methyl]-1H-imidazole-5-carboxylic Acid Ethyl Ester)
TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 4-(1-Hydroxy-1-methylethyl)-2-propyl-1-[[2'-[2-(triphenylmethyl)-2H-tetrazol-5-yl][1,1'-biphenyl]-4-yl]methyl]-1H-imidazole-5-carboxylic Acid Ethyl Ester
CAS 172875-59-1
Analyt- eller komponentnamn 4-(1-Hydroxy-1-methylethyl)-2-propyl-1-[[2'-[2-(triphenylmethyl)-2H-tetrazol-5-yl][1,1'-biphenyl]-4-yl]methyl]-1H-imidazole-5-carboxylic Acid Ethyl Ester
Frakt skick Room Temperature
Molekylformel C45 H44 N6 O3
Certifikat och efterlevnad ISO/IEC 17025
Synonym 1H-Imidazole-5-carboxylic acid, 4-(1-hydroxy-1-methylethyl)-2-propyl-1-[[2'-[2-(triphenylmethyl)-2H-tetrazol-5-yl][1,1'-biphenyl]-4-yl]methyl]-, ethyl ester, 4-(1-Hydroxy-1-methylethyl)-2-propyl-1-[[2'-[2-(triphenylmethyl)-2H-tetrazol-5-yl][1,1'-biphenyl]-4-yl]methyl]-1H-imidazole-5-carboxylic Acid Ethyl Ester
InChI formel InChI=1S/C45H44N6O3/c1-5-18-39-46-41(44(3,4)53)40(43(52)54-6-2)50(39)31-32-27-29-33(30-28-32)37-25-16-17-26-38(37)42-47-49-51(48-42)45(34-19-10-7-11-20-34,35-21-12-8-13-22-35)36-23-14-9-15-24-36/h7-17,19-30,53H,5-6,18,31H2,1-4H3
LEDER CCCc1nc(c(C(=O)OCC)n1Cc2ccc(cc2)c3ccccc3c4nnn(n4)C(c5ccccc5)(c6ccccc6)c7ccccc7)C(C)(C)O
IUPAC-namn ethyl 5-(2-hydroxypropan-2-yl)-2-propyl-3-[[4-[2-(2-trityltetrazol-5-yl)phenyl]phenyl]methyl]imidazole-4-carboxylate
Molekylvikt (g/mol) 716.87
Kvantitet 25 mg
Formel vikt 716.3475 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.