missing translation for 'onlineSavingsMsg'
Learn More

N1-(1-Oxo-2,3-dihydro-1H-inden-5-yl)acetamide, 97%, Thermo Scientific™

Catalog Number p-2705544
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
58161-35-6
C11H11NO2
189.214
MFCD00099465
GHUPGGYDRVSZSW-UHFFFAOYSA-N
n-1-oxo-2,3-dihydro-1h-inden-5-yl acetamide, n1-1-oxo-2,3-dihydro-1h-inden-5-yl acetamide, 5-acetamido-1-indanone, 5-acetylamino-1-indanone, n-1-oxoindan-5-yl acetamide, n-1-oxo-2,3-dihydroinden-5-yl acetamide, acetamide, n-2,3-dihydro-1-oxo-1h-inden-5-yl, 5-acetamido-2,3-dihydro-1-oxo-1h-indene, 5-acetamido-indanone
312894
N-(1-oxo-2,3-dihydroinden-5-yl)acetamide
CC(=O)NC1=CC2=C(C=C1)C(=O)CC2
This item is not returnable. View return policy
58161-35-6
C11H11NO2
189.214
MFCD00099465
GHUPGGYDRVSZSW-UHFFFAOYSA-N
n-1-oxo-2,3-dihydro-1h-inden-5-yl acetamide, n1-1-oxo-2,3-dihydro-1h-inden-5-yl acetamide, 5-acetamido-1-indanone, 5-acetylamino-1-indanone, n-1-oxoindan-5-yl acetamide, n-1-oxo-2,3-dihydroinden-5-yl acetamide, acetamide, n-2,3-dihydro-1-oxo-1h-inden-5-yl, 5-acetamido-2,3-dihydro-1-oxo-1h-indene, 5-acetamido-indanone
312894
N-(1-oxo-2,3-dihydroinden-5-yl)acetamide
CC(=O)NC1=CC2=C(C=C1)C(=O)CC2
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 189.21
Percent Purity 97%
Chemical Name or Material N1-(1-oxo-2, 3-dihydro-1H-inden-5-yl)acetamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'