Läs mer
N1-Ethylcarbamoyl Cabergoline, TRC
Produktkod. 17941465
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17941465 1 mg
17910895 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17941465 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE9009251MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 126554-50-5
Molekylformel C29 H42 N6 O3
Molekylvikt (g/mol) 522.68
Synonym Ergoline-1,8-dicarboxamide, N8-[3-(dimethylamino)propyl]-N1-ethyl-N8-[(ethylamino)carbonyl]-6-(2-propen-1-yl)-, (8β)-, (6aR,9R,10aR)-7-allyl-N9-[3-(dimethylamino)propyl]-N4-ethyl-N9-(ethylcarbamoyl)-6,6a,8,9,10,10a-hexahydroindolo[4,3-fg]quinoline-4,9-dicarboxamide, N1-Ethylcarbamoyl Cabergoline
IUPAC-namn (6aR,9R,10aR)-9-N-[3-(dimethylamino)propyl]-4-N-ethyl-9-N-(ethylcarbamoyl)-7-prop-2-enyl-6,6a,8,9,10,10a-hexahydroindolo[4,3-fg]quinoline-4,9-dicarboxamide
LEDER CCNC(=O)N(CCCN(C)C)C(=O)[C@@H]1C[C@H]2[C@@H](Cc3cn(C(=O)NCC)c4cccc2c34)N(CC=C)C1
Specifikationer
Kemiskt namn eller material N1-Ethylcarbamoyl Cabergoline
Kvantitet 1 mg
InChI formel InChI=1S/C29H42N6O3/c1-6-13-33-18-21(27(36)34(28(37)30-7-2)15-10-14-32(4)5)16-23-22-11-9-12-24-26(22)20(17-25(23)33)19-35(24)29(38)31-8-3/h6,9,11-12,19,21,23,25H,1,7-8,10,13-18H2,2-5H3,(H,30,37)(H,31,38)/t21-,23-,25-/m1/s1
Formel vikt 522.3318