Läs mer
N1,N12-Diacetylspermine Dihydrochloride, TRC
Produktkod. 30010169
Change view
Kvantitet:
2.5 mg
25 mg
Förpackningsstorlek:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30010169 2.5 mg 2.5mg
30006900 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30010169 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD3670052.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 77928-71-3
Molekylformel C14H32Cl2N4O2
Molekylvikt (g/mol) 359.34
Synonym N,N'-[1,4-butanediylbis(imino-3,1-propanediyl)]bis-acetamide Dihydrochloride, BAS, N,N'-[Tetramethylenebis(iminotrimethylene)]bis-acetamide Dihydrochloride, N1,N12-Bis(acetyl)spermine Dihydrochloride
LEDER Fc1cc2c(cc1)nc(o2)C3CCN(CC3)CCC4C(=O)N5C(CCCC5)=NC=4C
Specifikationer
Kemiskt namn eller material N1,N12-Diacetylspermine Dihydrochloride
Anteckningar om renhetsgrad HPLC
Kvantitet 2.5 mg
InChI formel InChI=1S/C14H30N4O2.2ClH/c1-13(19)17-11-5-9-15-7-3-4-8-16-10-6-12-18-14(2)20;;/h15-16H,3-12H2,1-2H3,(H,17,19)(H,18,20);2*1H
Formel vikt 358.1902
Procent renhet >95%