Läs mer
N1,N3-Bis[2-(3,4-dimethoxyphenyl)ethyl]-N1,N3-dimethyl-1,3-propanediamine Dihydrochloride, TRC
Produktkod. 30344163
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30344163 50 mg
1 options
Produktkod. 30344163 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB42991550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N,N'-Bis[2-(3,4-dimethoxyphenyl)ethyl]-N,N'-dimethylpropane-1,3-diamine Dihydrochloride
CAS 63434-11-7
Molekylformel C25 H38 N2 O4 . 2 Cl H
Kvantitet 50 mg
Synonym N,N'-Bis[2-(3,4-dimethoxyphenyl)ethyl]-N,N'-dimethylpropane-1,3-diamine Dihydrochloride, Verapamil Hydrochloride Imp. A (EP) as Dihydrochloride
InChI formel InChI=1S/C25H38N2O4.2ClH/c1-26(16-12-20-8-10-22(28-3)24(18-20)30-5)14-7-15-27(2)17-13-21-9-11-23(29-4)25(19-21)31-6;;/h8-11,18-19H,7,12-17H2,1-6H3;2*1H
LEDER Cl.Cl.COc1ccc(CCN(C)CCCN(C)CCc2ccc(OC)c(OC)c2)cc1OC
IUPAC-namn N,N'-bis[2-(3,4-dimethoxyphenyl)ethyl]-N,N'-dimethylpropane-1,3-diamine;dihydrochloride
Molekylvikt (g/mol) 503.5021
Formel vikt 502.2365