Läs mer
N1-Nornicergoline, TRC
Produktkod. 30134319
Change view
Kvantitet:
2.5 mg
5 mg
10 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30134319 5 mg -
30134051 2.5 mg 2.5mg
17913505 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134319 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN7566005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 35264-46-1
Molekylformel C23 H24 Br N3 O3
Molekylvikt (g/mol) 470.36
Synonym Ergoline-8-methanol, 10-methoxy-6-methyl-, 8-(5-bromo-3-pyridinecarboxylate), (8β)-, Ergoline-8-methanol, 10-methoxy-6-methyl-, 5-bromo-3-pyridinecarboxylate (ester), (8β)- (9CI), 1-Demethylnicergoline, N1-Demethylnicergoline, N1-Nornicergoline, [(6aR,9R,10aS)-10a-Methoxy7-methyl-4,6,6a,7,8,9,10,10a-octahydroindolo[4,3-fg]quinolin-9-yl]methyl 5-bromopyridine-3-carboxylate, 1-Desmethylnicergoline, Nicergoline Imp. B (EP)
IUPAC-namn [(6aR,9R,10aS)-10a-methoxy-7-methyl-4,6,6a,8,9,10-hexahydroindolo[4,3-fg]quinoline-9-yl]methyl 5-bromopyridine-3-carboxylate
LEDER CO[C@]12C[C@@H](COC(=O)c3cncc(Br)c3)CN(C)[C@@H]1Cc4c[nH]c5cccc2c45
Specifikationer
Kemiskt namn eller material 1-Demethylnicergoline
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C23H24BrN3O3/c1-27-12-14(13-30-22(28)16-6-17(24)11-25-9-16)8-23(29-2)18-4-3-5-19-21(18)15(10-26-19)7-20(23)27/h3-6,9-11,14,20,26H,7-8,12-13H2,1-2H3/t14-,20-,23+/m1/s1
Formel vikt 469.1001
Procent renhet >95%