Läs mer
N1,O2'-Dimethyladenosine, TRC
Produktkod. 30004841
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30004841 5 mg
30012717 1 mg
30012709 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30004841 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4474105MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 91101-00-7
Molekylformel C12H17N5O4
Molekylvikt (g/mol) 295.29
Synonym 1-Methyl-2'-O-methyladenosine, 2'-O-Methyl-1-methyladenosine;
LEDER O[C@H]1[C@@H](OC)[C@H](N2C(N=CN(C)C3=N)=C3N=C2)O[C@@H]1CO
Specifikationer
Kemiskt namn eller material N1,O2'-Dimethyladenosine
Kvantitet 5 mg
InChI formel InChI=1S/C12H19N5O4/c1-16-4-15-11-7(10(16)13)14-5-17(11)12-9(20-2)8(19)6(3-18)21-12/h5-6,8-9,12-13,15,18-19H,3-4H2,1-2H3/t6-,8-,9-,12-/m1/s1
Formel vikt 295.1281