Läs mer
N10-Monodesmethyl Rizatriptan, TRC
Produktkod. 30133798
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133798 1 mg
30012496 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30133798 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM5395001MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 144034-84-4
Molekylformel C14H17N5
Molekylvikt (g/mol) 255.32
Synonym N-Methyl-5-(1H-1,2,4-triazol-1-ylmethyl)-1H-indole-3-ethanamine, N-Methyl-2-[5-(1H-1,2,4-triazol-1-yl-methyl)-1H-indol-3-yl]ethanamine
LEDER CNCCc1c[nH]c2ccc(Cn3cncn3)cc12
Specifikationer
Kemiskt namn eller material N10-Monodesmethyl Rizatriptan
Kvantitet 1 mg
InChI formel InChI=1S/C14H17N5/c1-15-5-4-12-7-17-14-3-2-11(6-13(12)14)8-19-10-16-9-18-19/h2-3,6-7,9-10,15,17H,4-5,8H2,1H3