Läs mer
N2'-Deacetyl-N2'-[3-(methylthio)-1-oxopropyl]-maytansine, TRC
Produktkod. 30361888
Change view
Kvantitet:
1 mg
2.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30361888 1 mg
30357761 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30361888 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD1990351MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 912569-84-7
Molekylformel C36H50ClN3O10S
Molekylvikt (g/mol) 752.31
Synonym S-Me-DM1
IUPAC-namn (14S,16S,32S,33S,2R,4S,10E,12E,14R)-86-chloro-14-hydroxy-85,14-dimethoxy-33,2,7,10-tetramethyl-12,6-dioxo-7-aza-1(6,4)-oxazinana-3(2,3)-oxirana-8(1,3)-benzenacyclotetradecaphane-10,12-dien-4-yl N-methyl-N-(3-(methylthio)propanoyl)-L-alaninate
LEDER ClC1=C(N(C)C(C[C@@H]2OC([C@H](C)N(C)C(CCSC)=O)=O)=O)C=C(C=C1OC)C/C(C)=C/C=C/[C@@H](OC)[C@@]3(O)C[C@]([C@H]([C@H]4[C@]2(O4)C)C)([H])OC(N3)=O
Specifikationer
Kemiskt namn eller material N2'-Deacetyl-N2'-[3-(methylthio)-1-oxopropyl]-maytansine
Kvantitet 1 mg
InChI formel InChI=1S/C36H50ClN3O10S/c1-20-11-10-12-27(47-8)36(45)19-26(48-34(44)38-36)21(2)32-35(4,50-32)28(49-33(43)22(3)39(5)29(41)13-14-51-9)18-30(42)40(6)24-16-23(15-20)17-25(46-7)31(24)37/h10-12,16-17,21-22,26-28,32,45H,13-15,18-19H2,1-9H3,(H,38,44)/b12-10+,20-11+/t21-,22+,26+,27-,28+,32+,35+,36+/m1/s1
Formel vikt 751.29