Learn More
N2-Methyl Alfuzosin, TRC
Product Code. 30006932
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30006932 2.5 mg 2.5mg
30005729 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30006932 Supplier Toronto Research Chemicals Supplier No. TRCM2766502.5MG
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 72104-34-8
Molecular Formula C19 H27 N5 O4 . Cl H
Molecular Weight (g/mol) 425.91
Synonym Alfuzosin Hydrochloride Imp. C (EP) as Hydrochloride, (2RS)-N-[3-[(4-amino-6,7-dimethoxyquinazolin-2-yl)amino]propyl]-N-methyltetrahydrofuran-2-carboxamide Hydrochloride
IUPAC Name N-[3-[(4-amino-6,7-dimethoxyquinazolin-2-yl)amino]propyl]-N-methyloxolane-2-carboxamide;hydrochloride
SMILES Cl.COc1cc2nc(NCCCN(C)C(=O)C3CCCO3)nc(N)c2cc1OC
Specifications
Quantity 2.5 mg
InChI Formula InChI=1S/C19H27N5O4.ClH/c1-24(18(25)14-6-4-9-28-14)8-5-7-21-19-22-13-11-16(27-3)15(26-2)10-12(13)17(20)23-19;/h10-11,14H,4-9H2,1-3H3,(H3,20,21,22,23);1H
Formula Weight 425.183
Chemical Name or Material N2-Methyl Alfuzosin Hydrochloride