Läs mer
N2-Methyl Trimethoprim (Impurity), TRC
Produktkod. 30133300
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133300 100 mg
30012902 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30133300 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM331190100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 213745-86-9
Molekylformel C15 H20 N4 O3
Molekylvikt (g/mol) 304.34
Synonym N2-Methyl-5-(3,4,5-trimethoxybenzyl)pyrimidine-2,4-diamine, Trimethoprim Imp. A (EP)
IUPAC-namn 2-N-methyl-5-[(3,4,5-trimethoxyphenyl)methyl]pyrimidine-2,4-diamine
LEDER CNc1ncc(Cc2cc(OC)c(OC)c(OC)c2)c(N)n1
Specifikationer
Kemiskt namn eller material N2-Methyl-5-(3,4,5-trimethoxybenzyl)pyrimidine-2,4-diamine
Kvantitet 100 mg
InChI formel InChI=1S/C15H20N4O3/c1-17-15-18-8-10(14(16)19-15)5-9-6-11(20-2)13(22-4)12(7-9)21-3/h6-8H,5H2,1-4H3,(H3,16,17,18,19)
Formel vikt 304.1535