Läs mer
N2,N5-Dibenzoyl-4-oxo-L-ornithine Methyl Ester, TRC
Produktkod. 30367955
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30367955 25 mg
30356266 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30367955 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD41731525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 78420-25-4
Molekylformel C20H20N2O5
Molekylvikt (g/mol) 368.38
Synonym 2,5-Di(benzoylamino)-4-oxo-pentanoic Acid Methyl Ester
LEDER COC(=O)[C@H](CC(=O)CNC(=O)c1ccccc1)NC(=O)c2ccccc2
Specifikationer
Kemiskt namn eller material N2,N5-Dibenzoyl-4-oxo-L-ornithine Methyl Ester
Kvantitet 25 mg
InChI formel InChI=1S/C20H20N2O5/c1-27-20(26)17(22-19(25)15-10-6-3-7-11-15)12-16(23)13-21-18(24)14-8-4-2-5-9-14/h2-11,17H,12-13H2,1H3,(H,21,24)(H,22,25)/t17-/m0/s1
Formel vikt 368.1372