Läs mer
R&D Systems™ N21-MAX Vitamin A Free Media Supplement (50X)
:
N21-MAX Vitamin A free media supplement; 50X; Qty: 10mL
Beskrivning
- Supplied as 50X concentrated solution
- Limits retinoic acid production to reduce unwanted NSC differentiation
- Rigorous quality control testing ensures consistency across experiments
- Eliminates the need for an astrocyte feeder layer
- Used to mature and maintain healthy primary neurons in culture
- Quality tested to reduce unwanted experimental variation
- Contains factors qualified to the expansion of NSCs in vitro
- Does not require an astrocyte feeder layer
Specifikationer
Specifikationer
| Koncentration | 50X |
| Innehåll och lagring | Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Form | Liquid |
| Produkttyp | Media Supplement |
| Inkluderar | Albumin (bovine), L-Carnitine, Catalase, Corticosterone, Ethanolamine, Glutathione, Galactose, Holo-Transferrin, Insulin, Linoleic Acid, Linolenic Acid, Lipoic Acid, Progesterone, Putrescine, Selenite, Superoxide dismutase, Triiodo-L-thyronine, D,L-alpha-Tocopherol, D,L-alpha-Tocopherol acetate. Supplied in a volume sufficient to supplement 500mL of media at the recommended concentration. N21-MAX Vitamin A Free Media Supplement is tested for use in Neurobasal Media. |
| Kvantitet | 10 mL |
| Regulatorisk status | RUO |