Läs mer
N3-Methyl Esomeprazole, TRC
Produktkod. 30467916
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30467916 1 mg
1 options
Produktkod. 30467916 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM2457301MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N3-Methyl Esomeprazole
CAS 1346240-11-6
Molekylformel C18H21N3O3S
Kvantitet 1 mg
Synonym (S)-6-methoxy-2-(((4-methoxy-3,5-dimethylpyridin-2-yl)methyl)sulfinyl)-1-methyl-1H-benzo[d]imidazole
InChI formel InChI=1S/C18H21N3O3S/c1-11-9-19-15(12(2)17(11)24-5)10-25(22)18-20-14-7-6-13(23-4)8-16(14)21(18)3/h6-9H,10H2,1-5H3/t25-/m0/s1
LEDER O=[S@](C1=NC2=C(C=C(C=C2)OC)N1C)CC3=C(C(OC)=C(C=N3)C)C
Molekylvikt (g/mol) 359.44