Läs mer
N3PT, MedChemExpress
Produktkod. 16497086
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
Förpackningsstorlek:
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16497086 100 mg -
16497028 10 mg -
16459577 5 mg -
16413087 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16497086 Leverantör Medchem Express Leverantörsnummer HY16339
MedChemExpress N3PT(N3-pyridyl thiamine) is a potent and selective transketolase(TKT) inhibitor (IC50= 22 nM for Apo-TK) both in vitro and in vivo.
N3PT(N3-pyridyl thiamine) is a potent and selective transketolase(TKT) inhibitor (IC50= 22 nM for Apo-TK) both in vitro and in vivo.
Specifikationer
Kemiskt namn eller material N3PT
CAS 13860-66-7
Färg Light Yellow
Anteckningar om renhetsgrad Research
Molekylformel C13H19Cl2N3OS
Kvantitet 100 mg
Synonym N3-pyridyl thiamine
Löslighetsinformation DMSO : 2.86 mg/mL (8.50 mM; Need ultrasonic)
LEDER CC1=C(CCO)SC=[N+]1CC2=CC=C(C)N=C2N.[H]Cl.[Cl-]
Molekylvikt (g/mol) 336.28
Formel vikt 336.28
Procent renhet 98.98%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-programmed cell death