Läs mer
N4-Acetylcytidine, TRC
Produktkod. 30483549
Change view
Kvantitet:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30483549 10 g
30470188 5 g
30481002 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30483549 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN63304810G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 3768-18-1
Molekylformel C11H15N3O6
Molekylvikt (g/mol) 285.25
Synonym N-Acetylcytidine, 4-Acetylcytidine
IUPAC-namn N-[1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-2-oxopyrimidin-4-yl]acetamide
LEDER CC(=O)NC1=NC(=O)N(C=C1)[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O
Specifikationer
Kemiskt namn eller material N4-Acetylcytidine
Kvantitet 10 g
InChI formel InChI=1S/C11H15N3O6/c1-5(16)12-7-2-3-14(11(19)13-7)10-9(18)8(17)6(4-15)20-10/h2-3,6,8-10,15,17-18H,4H2,1H3,(H,12,13,16,19)/t6-,8-,9-,10-/m1/s1
Formel vikt 285.0961