Läs mer
N4-Isobutylquinoline-3,4-diamine Hydrochloride, Mikromol™
Produktkod. 30050140
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30050140 100 mg
30052668 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30050140 Leverantör Mikromol Leverantörsnummer MM1158.07
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N4-Isobutylquinoline-3,4-diamine Hydrochloride
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N4-Isobutylquinoline-3,4-diamine Hydrochloride
CAS 935521-01-0
Analyt- eller komponentnamn N4-Isobutylquinoline-3,4-diamine Hydrochloride
Frakt skick Room Temperature
Molekylformel C13 H17 N3 . Cl H
Certifikat och efterlevnad ISO 17025
Synonym N4-Isobutylquinoline-3,4-diamine Hydrochloride, 3,4-Quinolinediamine, N4-(2-methylpropyl)-, hydrochloride (1:1), Imiquimod Related Compound E as Hydrochloride
InChI formel InChI=1S/C13H17N3.ClH/c1-9(2)7-16-13-10-5-3-4-6-12(10)15-8-11(13)14;/h3-6,8-9H,7,14H2,1-2H3,(H,15,16);1H
LEDER Cl.CC(C)CNc1c(N)cnc2ccccc12
IUPAC-namn 4-N-(2-methylpropyl)quinoline-3,4-diamine;hydrochloride
Molekylvikt (g/mol) 251.76
Kvantitet 100 mg
Formel vikt 251.1189 g/mol