Läs mer
Invitrogen™ N4BP2L1 Polyclonal Antibody
Beskrivning
Immunogen sequence: SGKTTLARQL QHDFPRALIF STDDFFFRED GAYEFNPDFL EEAHEWNQ Antigen sequence identity: Mouse 90%; Rat 92%.
N4BP2L1 (NEDD4 Binding Protein 2 Like 1) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene includekinase activity. An important paralog of this gene isN4BP2.
Specifikationer
Specifikationer
| Antigen | N4BP2L1 |
| Användningsområden | Immunocytochemistry |
| Klassificering | Polyclonal |
| Koncentration | 0.3 mg/mL |
| Konjugera | Unconjugated |
| Formulering | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gen | N4BP2L1 |
| Genaccessionsnr. | Q5TBK1 |
| Gene Alias | 2410024N18Rik; AI428195; B230342M21Rik; CG018; CG081; hypothetical gene CG018; N4BP2L1; NEDD4 binding protein 2 like 1; NEDD4 binding protein 2-like 1; NEDD4-binding protein 2-like 1 |
| Gensymboler | N4BP2L1 |
| Visa mer |