missing translation for 'onlineSavingsMsg'
Läs mer

N6-{[(2-Phenylethyl)amino]carbonothioyl}lysine-13C6,15N2, TRC

Produktkod. 30422855
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30422855 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30422855 Supplier Toronto Research Chemicals Supplier No. TRCP3359191MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material N6-{[(2-Phenylethyl)amino]carbonothioyl}lysine-13C6,15N2
CAS 1609246-45-8
Molekylformel C913C6H23N15N2O2S
Kvantitet 1 mg
Synonym PEITC-[13C6,15N2]Lys
InChI formel InChI=1S/C15H23N3O2S/c16-13(14(19)20)8-4-5-10-17-15(21)18-11-9-12-6-2-1-3-7-12/h1-3,6-7,13H,4-5,8-11,16H2,(H,19,20)(H2,17,18,21)/t13-/m0/s1/i4+1,5+1,8+1,10+1,13+1,14+1,16+1,17+1
LEDER S=C([15NH][13CH2][13CH2][13CH2][13CH2][13C@H]([15NH2])[13C](O)=O)NCCC1=CC=CC=C1
Molekylvikt (g/mol) 317.37
Formel vikt 317.1653
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.