Läs mer
N6-Formyl-adenosine, TRC
Produktkod. 17969334
Change view
Kvantitet:
1 mg
2.5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17969334 1 mg
17959634 2.5 mg
17989834 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17969334 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF6934551MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6706-56-5
Molekylformel C11 H13 N5 O5
Molekylvikt (g/mol) 295.251
Synonym N-Formyl-adenosine, N-(9-β-D-Ribofuranosyl-9H-purin-6-yl)formamide;
IUPAC-namn N-[9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl]purin-6-yl]formamide
LEDER OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)n2cnc3c(NC=O)ncnc23
Specifikationer
Kemiskt namn eller material N6-Formyl-adenosine
Kvantitet 1 mg
InChI formel InChI=1S/C11H13N5O5/c17-1-5-7(19)8(20)11(21-5)16-3-14-6-9(15-4-18)12-2-13-10(6)16/h2-5,7-8,11,17,19-20H,1H2,(H,12,13,15,18)/t5-,7-,8-,11-/m1/s1
Formel vikt 295.0917