Läs mer
N6-Methyladenosine-5'-monophosphate, TRC
Produktkod. 30482847
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30482847 10 mg
1 options
Produktkod. 30482847 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM27589810MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N6-Methyladenosine-5'-monophosphate Sodium Salt
CAS 4229-50-9
Molekylformel C11H16N5O7P
Kvantitet 10 mg
Synonym 5′-Adenylic acid','N-methyl- (9CI','ACI), Adenine','N6-methyl-9-β-D-ribofuranosyl-','5′-phosphate (6CI), Adenosine','N-methyl-','5′-(dihydrogen phosphate) (8CI), Adenosine','N-methyl-','5′-phosphate (7CI), N-Methyl-5′-adenylic acid (ACI), 6-Methyladenosine 5′-monophosphate, 6-Methylaminopurine ribotide, N6-Methyl-5′-AMP, N6-Methyl-AMP, N6-Methyladenosine 5′-monophosphate, N6-Methyladenosine 5′-phosphate, N6-Methyladenylic acid
InChI formel InChI=1S/C11H16N5O7P/c1-12-9-6-10(14-3-13-9)16(4-15-6)11-8(18)7(17)5(23-11)2-22-24(19,20)21/h3-5,7-8,11,17-18H,2H2,1H3,(H,12,13,14)(H2,19,20,21)/t5-,7-,8-,11-/m1/s1
LEDER CNC1=NC=NC2=C1N=CN2[C@H]3[C@@H]([C@@H]([C@H](O3)COP(O)(O)=O)O)O
IUPAC-namn ((2R,3S,4R,5R)-3,4-dihydroxy-5-(6-(methylamino)-9H-purin-9-yl)tetrahydrofuran-2-yl)methyl dihydrogen phosphate
Molekylvikt (g/mol) 361.25
Formel vikt 361.08