Läs mer
N6-Methyladenosine, TRC
Produktkod. 30467859
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30467859 500 mg
1 options
Produktkod. 30467859 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM275895500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N6-Methyladenosine
CAS 1867-73-8
Molekylformel C11H15N5O4
Kvantitet 500 mg
Synonym 6-Methyladenosine, N-methyladenosine, 6-Methylamino-9-β-D-ribofuranosylpurine, 6-Methylaminopurine D-riboside, 6-Methylaminopurine ribonucleoside, 6-Methylaminopurine riboside, 9-β-D-Ribofuranosyl-N6-methylaminopurine, N-Methyladenosine, N6-Methyladenosine, NSC 29409
InChI formel InChI=1S/C11H15N5O4/c1-12-9-6-10(14-3-13-9)16(4-15-6)11-8(19)7(18)5(2-17)20-11/h3-5,7-8,11,17-19H,2H2,1H3,(H,12,13,14)/t5-,7-,8-,11-/m1/s1
LEDER OC[C@@H]1[C@@H](O)[C@@H](O)[C@H](N2C(N=CN=C3NC)=C3N=C2)O1
Molekylvikt (g/mol) 281.27
Formel vikt 281.1124