Läs mer
N6-(N-Threonylcarbonyl)adenosine-13C4,15N, TRC
Produktkod. 30454185
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30454185 1 mg
30454001 10 mg
30453297 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30454185 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT4055621MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1195030-28-4
Molekylformel C1113C4H20N515NO8
Molekylvikt (g/mol) 417.32
Synonym N-[[(9-beta-D-Ribofuranosyl-9H-purin-6-yl)amino]carbonyl]-L-threonine-1,2,3,4-13C4-15N, N-(6-Nebularinylcarbamoyl)threonine-13C4,15N, N-(6-Purinylcarbamoyl)-L-threonine Riboside-13C4,15N, N-[(9-beta-D-Ribofuranosyl-9H-purin-6-yl)carbamoyl]-L-threonine-13C4,15N, N-[(9-beta-D-Ribofuranosyl-9H-purin-6-yl)carbamoyl]threonine-13C4,15N, N-[(9-beta-D-Ribofuranosylpurin-6-yl)carbamoyl]threonine-13C4,15N, N-[N-(9-beta-D-Ribofuranosylpurin-6-yl)carbamoyl]threonine-13C4,15N, N-[[(9-beta-D-Ribofuranosyl-9H-purin-6-yl)amino]carbonyl]-L-threonine-13C4,15N, N6-Threonylcarbamoyladenosine-13C4,15N, t6-Adenosine-13C4,15N
IUPAC-namn (2S,3R)-2-[[9-[(2R,3S,4R,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]purin-6-yl]carbamoylamino]-3-hydroxy(1,2,3,4-^{13}C_{4})butanoic acid
LEDER [13CH3][13C@@H](O)[13C@H]([15NH]C(=O)Nc1ncnc2c1ncn2[C@@H]3O[C@H](CO)[C@H](O)[C@@H]3O)[13C](=O)O
Specifikationer
Kemiskt namn eller material N6-(N-Threonylcarbonyl)adenosine-13C4,15N
Kvantitet 1 mg
InChI formel InChI=1S/C15H20N6O8/c1-5(23)7(14(26)27)19-15(28)20-11-8-12(17-3-16-11)21(4-18-8)13-10(25)9(24)6(2-22)29-13/h3-7,9-10,13,22-25H,2H2,1H3,(H,26,27)(H2,16,17,19,20,28)/t5-,6-,7+,9+,10+,13-/m1/s1/i1+1,5+1,7+1,14+1,19+1
Formel vikt 417.1447