Läs mer
N7-(2-Carbamoyl-2-hydroxyethyl)guanine, TRC
Produktkod. 30356430
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30356430 5 mg
30356558 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30356430 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC1758755MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 163734-06-3
Molekylformel C8 H10 N6 O3
Molekylvikt (g/mol) 238.2
IUPAC-namn 3-(2-amino-6-oxo-1H-purin-7-yl)-2-hydroxypropanamide
LEDER NC(=O)C(O)Cn1cnc2N=C(N)NC(=O)c12
Specifikationer
Kemiskt namn eller material N7-(2-Carbamoyl-2-hydroxyethyl)guanine
Kvantitet 5 mg
InChI formel InChI=1S/C8H10N6O3/c9-5(16)3(15)1-14-2-11-6-4(14)7(17)13-8(10)12-6/h2-3,15H,1H2,(H2,9,16)(H3,10,12,13,17)
Formel vikt 238.0814