Läs mer
N8-Desethylcarbamoyl-N1-ethylcarbamoyl Cabergoline (Cabergoline EP Impurity B), TRC
Produktkod. 17960075
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17960075 1 mg
17908014 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17960075 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2895451MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 166533-36-4
Molekylformel C26 H37 N5 O2
Molekylvikt (g/mol) 451.6
Synonym Ergoline-1,8-dicarboxamide, N8-[3-(dimethylamino)propyl]-N1-ethyl-6-(2-propen-1-yl)-, (8β)-, (6aR,9R,10aR)-7-Allyl-N9-[3-(dimethylamino)propyl]-N4-ethyl-6,6a,8,9,10,10a-hexahydroindolo[4,3-fg]quinoline-4,9-dicarboxamide
IUPAC-namn (6aR,9R,10aR)-9-N-[3-(dimethylamino)propyl]-4-N-ethyl-7-prop-2-enyl-6,6a,8,9,10,10a-hexahydroindolo[4,3-fg]quinoline-4,9-dicarboxamide
LEDER CCNC(=O)n1cc2C[C@@H]3[C@H](C[C@H](CN3CC=C)C(=O)NCCCN(C)C)c4cccc1c24
Specifikationer
Kemiskt namn eller material N8-Desethylcarbamoyl-N1-ethylcarbamoyl Cabergoline
Kvantitet 1 mg
InChI formel InChI=1S/C26H37N5O2/c1-5-12-30-16-19(25(32)28-11-8-13-29(3)4)14-21-20-9-7-10-22-24(20)18(15-23(21)30)17-31(22)26(33)27-6-2/h5,7,9-10,17,19,21,23H,1,6,8,11-16H2,2-4H3,(H,27,33)(H,28,32)/t19-,21-,23-/m1/s1
Formel vikt 451.2947