Läs mer
Nabumetone, TRC
Produktkod. 30466021
Change view
Kvantitet:
50 mg
10 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30466021 100 g
30473677 50 mg
30481562 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30466021 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN200500100G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 42924-53-8
Molekylformel C15 H16 O2
Molekylvikt (g/mol) 228.29
Synonym Nabumetone, 4-(6-Methoxy-2-naphthalenyl)-2-butanone, 1-(2'-Methoxynaphth-6'-yl)butan-3-one, 4-(6-Methoxy-2-naphthyl)-2-butanone, Arthaxan, BRL 14777, Balmox, Consolan, Nabumeton, Nabumetone, Nabuser, Relafen, Relifen, Relifex
IUPAC-namn 4-(6-methoxynaphthalen-2-yl)butan-2-one
LEDER COc1ccc2cc(CCC(=O)C)ccc2c1
Specifikationer
Kemiskt namn eller material Nabumetone
Kvantitet 100 g
InChI formel InChI=1S/C15H16O2/c1-11(16)3-4-12-5-6-14-10-15(17-2)8-7-13(14)9-12/h5-10H,3-4H2,1-2H3
Formel vikt 228.115