Läs mer
Nadoxolol Hydrochloride, TRC
Produktkod. 30132804
Change view
Kvantitet:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30132804 25 mg
17920635 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30132804 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN20104025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 35991-93-6
Molekylformel C14H17ClN2O3
Molekylvikt (g/mol) 296.75
Synonym N,3-Dihydroxy-4-(1-naphthalenyloxy)-butanimidamide Monohydrochloride, 4-(1-Naphthoxy)-3-hydroxybutyramidoxime hydrochloride, Bradyl, LL 1530
IUPAC-namn N,3-dihydroxy-4-naphthalen-1-yloxybutanimidamide;hydrochloride
LEDER OC(CC(NO)=N)COC1=C2C(C=CC=C2)=CC=C1.Cl
Specifikationer
Kemiskt namn eller material Nadoxolol Hydrochloride
Kvantitet 25 mg
InChI formel InChI=1S/C14H16N2O3.ClH/c15-14(16-18)8-11(17)9-19-13-7-3-5-10-4-1-2-6-12(10)13;/h1-7,11,17-18H,8-9H2,(H2,15,16);1H
Formel vikt 296.0928