Läs mer
Nafcillin Penilloic Acid (Mixture of Diastereomers), TRC
Produktkod. 30474391
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30474391 1 mg
1 options
Produktkod. 30474391 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN2115151MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Nafcillin Penilloic Acid (Mixture of Diastereomers)
CAS 940306-31-0
Molekylformel C21 H24 N2 O6 S
Kvantitet 1 mg
Synonym (αR,4S)-4-Carboxy-α-[[(2-ethoxy-1-naphthalenyl)carbonyl]amino]-5,5-dimethyl-2-thiazolidineacetic Acid
InChI formel InChI=1S/C21H24N2O6S/c1-4-29-13-10-9-11-7-5-6-8-12(11)14(13)17(24)22-15(19(25)26)18-23-16(20(27)28)21(2,3)30-18/h5-10,15-16,18,23H,4H2,1-3H3,(H,22,24)(H,25,26)(H,27,28)/t15-,16-,18?/m0/s1
LEDER CCOc1ccc2ccccc2c1C(=O)N[C@@H](C3N[C@@H](C(=O)O)C(C)(C)S3)C(=O)O
IUPAC-namn (4S)-2-[(R)-carboxy-[(2-ethoxynaphthalene-1-carbonyl)amino]methyl]-5,5-dimethyl-thiazolidine-4-carboxylic acid
Molekylvikt (g/mol) 432.49
Formel vikt 432.1355