Läs mer
Naftidrofuryl Hydrogen Oxalate, LoGiCal
Produktkod. 30462889
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30462889 10 mg
1 options
Produktkod. 30462889 Leverantör LoGiCal Leverantörsnummer LGCFOR0373.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Naftidrofuryl Hydrogen Oxalate
CAS 6-4-3200
Analyt- eller komponentnamn Naftidrofuryl Hydrogen Oxalate
Frakt skick Room Temperature
Molekylformel C24 H33 N O3 . C2 H2 O4
Certifikat och efterlevnad ISO 9001
Synonym Naftidrofuryl Hydrogen Oxalate, Citoxid, Di-Actane, Dubimax, Dusodril, EU 1806, Iridus, LS 121, NSC 225233, Nafronyl acid oxalate, Nafronyl oxalate, Naftidrofuryl oxalate, Naphtidrofuryl oxalate, Praxilene, 2-Furanpropanoic acid, tetrahydro-alpha-(1-naphthalenylmethyl)-, 2-(diethylamino)ethyl ester, ethanedioate (1:1), 2-Furanpropionic acid, tetrahydro-alpha-(1-naphthylmethyl)-, 2-(diethylamino)ethyl ester oxalate (1:1) (8CI), 2-Furanpropionic acid, tetrahydro-α-(1-naphthylmethyl)-, 2-(diethylamino)ethyl ester oxalate (1:1) (8CI)
InChI formel InChI=1S/C24H33NO3.C2H2O4/c1-3-25(4-2)14-16-28-24(26)21(18-22-12-8-15-27-22)17-20-11-7-10-19-9-5-6-13-23(19)20;3-1(4)2(5)6/h5-7,9-11,13,21-22H,3-4,8,12,14-18H2,1-2H3;(H,3,4)(H,5,6)
Lösningstyp Neat
LEDER CCN(CC)CCOC(=O)C(CC1CCCO1)Cc2cccc3ccccc23.OC(=O)C(=O)O
IUPAC-namn 2-(diethylamino)ethyl 2-(naphthalen-1-ylmethyl)-3-(oxolan-2-yl)propanoate;oxalic acid
Industry Type Clinical Testing
Molekylvikt (g/mol) 473.56
Kvantitet 10 mg
Formel vikt 473.2414 g/mol